Läs mer
L-Alanine 2-Ethylbutyl Ester Hydrochloride, TRC
Produktkod. 30322657
Change view
Kvantitet:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30322657 5 g
30320865 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30322657 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1952205G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-Alanine 2-Ethylbutyl Ester Hydrochloride
CAS 946511-97-3
Molekylformel C9 H19 N O2 . Cl H
Kvantitet 5 g
Synonym L-Alanine, 2-ethylbutyl ester, hydrochloride (1:1), L-Alanine 2-Ethylbutyl Ester Hydrochloride
InChI formel InChI=1S/C9H19NO2.ClH/c1-4-8(5-2)6-12-9(11)7(3)10;/h7-8H,4-6,10H2,1-3H3;1H/t7-;/m0./s1
LEDER Cl.CCC(CC)COC(=O)[C@H](C)N
IUPAC-namn 2-ethylbutyl (2S)-2-aminopropanoate;hydrochloride
Molekylvikt (g/mol) 209.7136
Formel vikt 209.1183