Läs mer
L-allo-Threonine, 99%
Produktkod. p-7029796
Change view
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
15463657 250 mg 250mg
11352369 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 15463657 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H27301.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
CAS: 28954-12-3 | C4H9NO3 | 119.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28954-12-3
Molekylformel C4H9NO3
Molekylvikt (g/mol) 119.12
MDL-nummer MFCD00064268
InChI-nyckel AYFVYJQAPQTCCC-HRFVKAFMSA-N
Synonym l-allo-threonine, l-allothreonine, 2s,3s-2-amino-3-hydroxybutanoic acid, l +-allo-threonine, allo-l-threonine, h-allo-thr-oh, allothreonine, allo-threonine, unii-hcq253ckvk, dl-allo-threonine
PubChem CID 99289
ChEBI CHEBI:28718
IUPAC-namn (2S,3S)-2-amino-3-hydroxybutanoic acid
LEDER C[C@H](O)[C@H](N)C(O)=O
Specifikationer
Kemiskt namn eller material L-allo-Threonine
Smältpunkt 272°C (decomposition)
Beilstein 1721645
Kvantitet 250 mg
Formel vikt 119.12
Optisk rotation +9.2° (c=2 in Water)
Procent renhet 99%