Läs mer
L-Allooctopine, TRC
Produktkod. 30322494
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30322494 250 mg
1 options
Produktkod. 30322494 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA546800250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-Allooctopine
CAS 63358-47-4
Molekylformel C9H18N4O4
Kvantitet 250 mg
Synonym L-(+)-Allooctopine, N2-[(1S)-1-Carboxyethyl]-L-arginine, N2-(1-Carboxyethyl)-L-arginine
LEDER NC(=N)NCCC[C@H](NCCC(=O)O)C(=O)O
Molekylvikt (g/mol) 246.46
Formel vikt 246.1328