Läs mer
L-alpha-Aminosuberic Acid, TRC
Produktkod. 30317529
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30317529 50 mg
1 options
Produktkod. 30317529 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA61907550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-alpha-Aminosuberic Acid
CAS 4254-88-0
Molekylformel C8 H15 N O4
Kvantitet 50 mg
Synonym (S)-2-Aminooctanedioic Acid, L-2-Amino-octanedioic Acid (2S)-2-Aminooctanedioic Acid, L-2-Aminosuberic Acid, L-α-Aminosuberic Acid
InChI formel InChI=1S/C8H15NO4/c9-6(8(12)13)4-2-1-3-5-7(10)11/h6H,1-5,9H2,(H,10,11)(H,12,13)/t6-/m0/s1
LEDER N[C@@H](CCCCCC(=O)O)C(=O)O
IUPAC-namn (2S)-2-aminooctanedioic acid
Molekylvikt (g/mol) 189.209
Formel vikt 189.1