Läs mer
L-Aminobutyric Acid-d3, TRC
Produktkod. 30327201
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30327201 50 mg
1 options
Produktkod. 30327201 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60293250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-Aminobutyric Acid-d3
CAS 929202-07-3
Molekylformel C4 2H3 H6 N O2
Kvantitet 50 mg
Synonym Butanoic-4,4,4-d3 acid, 2-amino-, (2S)-, (2S)-2-Aminobutanoic-4,4,4-d3 acid, L-2-Aminobutyric acid-d3, L-Butyrine-d3, L-Ethylglycine-d3, L-Homoalanine-d3, L-α-Aminobutyric acid-d3, (S)-2-Aminobutanoic acid-d3, (S)-2-Aminobutanoic-4,4,4-D3 acid
InChI formel InChI=1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m0/s1/i1D3
LEDER [2H]C([2H])([2H])C[C@H](N)C(=O)O
IUPAC-namn (2S)-2-amino-4,4,4-trideuteriobutanoic acid
Molekylvikt (g/mol) 106.14
Formel vikt 106.0822