Läs mer
L-Arabinitol, TRC
Produktkod. 17939964
Change view
Kvantitet:
2.5 g
5 g
10 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17939964 2.5 g 2.5g
17948654 10 g -
17929824 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17939964 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7356102.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7643-75-6
Molekylformel C5 H12 O5
Molekylvikt (g/mol) 152.15
Synonym Arabinitol, L- (8CI), (-)-Arabitol, L-(-)-Arabitol, L-Arabitol, L-Lyxitol, L-arabino-Pentitol, NSC 83253
IUPAC-namn (2S,4S)-pentane-1,2,3,4,5-pentol
LEDER OC[C@H](O)C(O)[C@@H](O)CO
Specifikationer
Kemiskt namn eller material L-(-)-Arabitol
Kvantitet 2.5 g
InChI formel InChI=1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4-/m0/s1
Formel vikt 152.0685