Läs mer
L-Arginine-13C6 Hydrochloride, TRC
Produktkod. 30007869
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007869 5 mg
30006668 1 mg
17970045 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007869 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7695075MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 201740-91-2
Molekylformel 13C6 H14 N4 O2 . Cl H
Molekylvikt (g/mol) 216.6178
Synonym L-Arginine-guanidino,1,2,3,4,5-13C6, hydrochloride (1:1), L-Arginine-13C6, monohydrochloride (9CI), L-Arginine-13C6 Monohydrochloride, L-Arginine-guanidino,1,2,3,4,5-13C6 hydrochloride (1:1), (2S)-2-Amino-5-[(diaminomethylidene-13C)amino]pentanoic-1,2,3,4,5-13C5 acid hydrochloride, (2S)-2-Amino-5-carbamimidamidopentanoic-13C6 acid hydrochloride, (S)-2-Amino-5-(guanidino-13C)pentanoic-1,2,3,4,5-13C5 acid hydrochloride, (S)-Arginine-13C6 hydrochloride, 1,2,3,4,5-13C6 L-Arginine-guanidino Hydrochloride
IUPAC-namn (2S)-2-amino-5-[(C-aminocarbonimidoyl)amino](1,2,3,4,5-^{13}C_{5})pentanoic acid;hydrochloride
LEDER Cl.N[13C@@H]([13CH2][13CH2][13CH2]N[13C](=N)N)[13C](=O)O
Specifikationer
Kemiskt namn eller material L-Arginine-13C6 Hydrochloride
Kvantitet 5 mg
InChI formel InChI=1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1/i1+1,2+1,3+1,4+1,5+1,6+1;
Formel vikt 216.1085