Läs mer
L-Arginine-15N4 Hydrochloride, TRC
Produktkod. 30330346
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30330346 10 mg
1 options
Produktkod. 30330346 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA76950210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-Arginine-15N4 Hydrochloride
CAS 204633-95-4
Molekylformel C6 H14 15N4 O2 . Cl H
Kvantitet 10 mg
Synonym L-Arginine-guanidino,guanidino,guanidino,N2-15N4, hydrochloride (1:1), L-Arginine-15N4, monohydrochloride (9CI), L-Arginine-15N4 hydrochloride, (2S)-2-(Amino-15N)-5-[(diaminomethylidene-15N2)amino-15N]pentanoic acid hydrochloride, (2S)-2-Amino-5-carbamimidamidopentanoic-15N4 acid hydrochloride, (S)-2-(Amino-15N)-5-(guanidino-15N3)pentanoic acid hydrochloride, (S)-Arginine-15N4 hydrochloride
InChI formel InChI=1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1/i7+1,8+1,9+1,10+1;
LEDER Cl.[15NH2][C@@H](CCC[15NH]C(=[15NH])[15NH2])C(=O)O
IUPAC-namn (2S)-2-(^{15}N)azanyl-5-[(C-(^{15}N)azanylcarbonimidoyl)amino]pentanoic acid;hydrochloride
Molekylvikt (g/mol) 214.64
Formel vikt 214.0765