missing translation for 'onlineSavingsMsg'
Läs mer

L-Arginine-2,3,3,4,4,5,5-d7 HCl, TRC

Produktkod. 30333530
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30333530 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30333530 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCA76950110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material L-Arginine d7 hydrochloride
CAS 204244-77-9
Anteckningar om renhetsgrad HPLC
Molekylformel C6 D7 H7 N4 O2 . H Cl
Kvantitet 10 mg
Synonym L-Arginine-2,3,3,4,4,5,5-d7, hydrochloride (1:1), L-Arginine-2,3,3,4,4,5,5-d7, monohydrochloride (9CI), L-Arginine-2,3,3,4,4,5,5-d7 hydrochloride (1:1), L-Arginine-d7 hydrochloride, L-Arginine-2,3,3,4,4,5,5-D7 hydrochloride, (2S)-2-Amino-5-[(diaminomethylidene)amino]pentanoic-2,3,3,4,4,5,5-d7 acid hydrochloride, (2S)-2-Amino-5-carbamimidamidopentanoic-d7 acid hydrochloride, (S)-2-Amino-5-guanidinopentanoic-2,3,3,4,4,5,5-d7 acid hydrochloride, (S)-Arginine-d7 hydrochloride
InChI formel InChI=1S/C6H14N4O2.ClH/c7-4(5(11)12)2-1-3-10-6(8)9;/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);1H/t4-;/m0./s1/i1D2,2D2,3D2,4D;
LEDER Cl.[2H]C([2H])(NC(=N)N)C([2H])([2H])C([2H])([2H])[C@]([2H])(N)C(=O)O
IUPAC-namn (2S)-2-amino-2,3,3,4,4,5,5-heptadeuterio-5-guanidino-pentanoic acid;hydrochloride
Molekylvikt (g/mol) 217.7
Formel vikt 217.1323
Procent renhet >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.