Läs mer
L-Arginine-[4-nitro-3-(isopropylcarboxamido)phenyl] amide dihydrochloride, TRC
Produktkod. 30341898
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30341898 10 mg
1 options
Produktkod. 30341898 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA79496310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-Arginine-[4-nitro-3-(isopropylcarboxamido)phenyl] amide dihydrochloride
CAS 1272755-10-8
Molekylformel C16H25N7O4.2[HCl]
Kvantitet 10 mg
Synonym Arginine-5-Amino-2-Nitrobenzoic Acid Isopropylamide Dihydrochloride
InChI formel InChI=1S/C16H25N7O4.2ClH/c1-9(2)21-14(24)11-8-10(5-6-13(11)23(26)27)22-15(25)12(17)4-3-7-20-16(18)19;;/h5-6,8-9,12H,3-4,7,17H2,1-2H3,(H,21,24)(H,22,25)(H4,18,19,20);2*1H/t12-;;/m0../s1
LEDER Cl.Cl.CC(C)NC(=O)c1cc(NC(=O)[C@@H](N)CCCN=C(N)N)ccc1[N+](=O)[O-]
IUPAC-namn 5-[[(2S)-2-amino-5-(diaminomethylideneamino)pentanoyl]amino]-2-nitro-N-propan-2-ylbenzamide;dihydrochloride
Molekylvikt (g/mol) 452.33
Formel vikt 451.1502