missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Arginine hydrochloride, 98.5 to 101.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic Bottle
Specifications
Melting Point | 216°C (decomposition) |
Linear Formula | NH2C(NH)NH(CH2)3CH(NH2)COOH.HCl |
Beilstein | 3631658 |
Packaging | Plastic Bottle |
Formula Weight | 210.66 |
Percent Purity | 98.5 to 101.5% |
Density | 1.42 |
Quantity | 500 g |
Specific Gravity | 1.42 |
Physical Form | Crystalline Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'