Läs mer
L-Arginine L-Aspartate, TRC
Produktkod. 30330279
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30330279 1 g
1 options
Produktkod. 30330279 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7949201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Arginine aspartate
CAS 7675-83-4
Anteckningar om renhetsgrad HPLC
Molekylformel C6 H14 N4 O2 . C4 H7 N O4
Kvantitet 1 g
Synonym Arginine aspartate, (2S)-2-Amino-5-guanidinopentanoic acid (2S)-2-aminobutanedioate, L-Aspartic acid, compd. with L-arginine (1:1), Aspartic acid, L-, compd. with L-arginine (1:1) (8CI), Arginine, L-, L-aspartate (1:1) (8CI), L-Arginine, L-aspartate (1:1), Arginine aspartate
InChI formel InChI=1S/C6H14N4O2.C4H7NO4/c7-4(5(11)12)2-1-3-10-6(8)9;5-2(4(8)9)1-3(6)7/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);2H,1,5H2,(H,6,7)(H,8,9)/t4-;2-/m00/s1
LEDER N[C@@H](CCCNC(=N)N)C(=O)O.N[C@@H](CC(=O)O)C(=O)O
IUPAC-namn (2S)-2-aminobutanedioic acid;(2S)-2-amino-5-carbamimidamidopentanoic acid
Molekylvikt (g/mol) 307.3
Formel vikt 307.1492