Läs mer
L-Arginine L-Glutamate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | L-Arginine L-Glutamate |
| CAS | 4320-30-3 |
| Molekylformel | C6H14N4O2.C5H9NO4 |
| Kvantitet | 100 mg |
| Synonym | (S)-2-Amino-5-guanidinopentanoic Acid Compound With (S)-2-Aminopentanedioic Acid (1:1), L-Arginine, L-glutamate (1:1), Argimate, Arginine Glutamate, Ginamate, Glutamic Acid Compd. With Arginine, Glutargin, Glutepar, Modumate, NSC 122009, NSC 164937 |
| InChI formel | InChI=1S/C6H14N4O2.C5H9NO4/c7-4(5(11)12)2-1-3-10-6(8)9;6-3(5(9)10)1-2-4(7)8/h4H,1-3,7H2,(H,11,12)(H4,8,9,10);3H,1-2,6H2,(H,7,8)(H,9,10)/t4-;3-/m00/s1 |
| LEDER | N[C@@H](CCCNC(=N)N)C(=O)O.N[C@@H](CCC(=O)O)C(=O)O |
| IUPAC-namn | (2S)-2-amino-5-(diaminomethylideneamino)pentanoic acid;(2S)-2-aminopentanedioic acid |
| Molekylvikt (g/mol) | 321.33 |
| Formel vikt | 321.1648 |