missing translation for 'onlineSavingsMsg'
Läs mer

l-Arginine, Millipore™

Produktkod. 10688371
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 g
1 kg
10 kg
Förpackningsstorlek:
100g
10kg
1kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
10688371 1 kg 1kg
10265250 100 g 100g
10547782 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 10688371 Leverantör Millipore Leverantörsnummer 1015421000
 Begränsad artikel
Beräknad leverans: 27-01-2027
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

For biochemistry

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-79-3

Specifications

Kemiskt namn eller material L-Arginine
Smältpunkt 238°C (decomposition)
MDL-nummer MFCD00002635
Synonym L-Arginine, Arg, L-Arginine
Molekylvikt (g/mol) 174.2
Kvantitet 1 kg
Kvalitet Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.