Läs mer
L-Argininic Acid, TRC
Produktkod. 17974845
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17974845 50 mg
17961605 25 mg
17971925 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17974845 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA76919550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 157-07-3
Molekylformel C6 H13 N3 O3
Molekylvikt (g/mol) 175.19
Synonym Argininic Acid, (2S)-5-[(Aminoiminomethyl)amino]-2-hydroxypentanoic Acid, L-5-Guanidino-2-hydroxyvaleric Acid, (S)-5-[(Aminoiminomethyl)amino]-2-hydroxy-pentanoic Acid, (S)-5-Guanidino-2-hydroxypentanoicacid
IUPAC-namn (2S)-5-guanidino-2-hydroxy-pentanoic acid
LEDER NC(=N)NCCC[C@H](O)C(=O)O
Specifikationer
Kemiskt namn eller material L-Argininic Acid
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C6H13N3O3/c7-6(8)9-3-1-2-4(10)5(11)12/h4,10H,1-3H2,(H,11,12)(H4,7,8,9)/t4-/m0/s1
Kvantitet 50 mg
Formel vikt 175.0957
Procent renhet >95%