Läs mer
L-Ascorbic Acid-13C6, TRC
Produktkod. 30007470
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30007470 1 mg -
30004098 2.5 mg 2.5mg
30006910 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007470 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7869921MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1331939-77-5
Molekylformel 13C6H8O6
Molekylvikt (g/mol) 182.08
Synonym (+)-Ascorbic Acid-13C6, 100M-13C6, 3-Oxo-L-gulofuranolactone-13C6, 3-keto-L-Gulofuranolactone-13C6, Adenex-13C6, Allercorb-13C6, Ascorbajen-13C6, Cantan-13C6, Cantaxin-13C6, Vitacimin-13C6, Davitamon C-13C6, Laroscorbine-13C6, Vitamisin-13C6, Vitascorbol-13C6;
IUPAC-namn (2R)-2-(1,2-dihydroxy(1,2-13C2)ethyl)-3,4-dihydroxy-(2,3,4,5-13C4)2H-furan-5-one
LEDER O[13C]([13C@H]1[13C@@H](O)[13CH2]O)=[13C](O)[13C](O1)=O
Specifikationer
Kemiskt namn eller material L-Ascorbic Acid-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2?,5-/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1
Procent renhet >95%