Läs mer
L-Ascorbic acid 2-(Dihydrogen Phosphate) Magnesium Salt (Technical Grade), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | L-Ascorbic acid 2-(Dihydrogen Phosphate) Magnesium Salt |
| CAS | 113170-55-1 |
| Molekylformel | C6H9O9P xMg |
| Kvantitet | 1 g |
| Synonym | Ascorbic acid phosphate dimagnesium salt, Ascorbyl PM, Magnesium ascorbyl phosphate, Phospitan C |
| InChI formel | InChI=1S/C6H9O9P.Mg/c7-1-2(8)4-3(9)5(6(10)14-4)15-16(11,12)13;/h2,4,7-9H,1H2,(H2,11,12,13);/q;+2/p-2/t2-,4+;/m0./s1 |
| LEDER | CCOc1c(cccc1)O[C@@H](c2ccccc2)[C@@H](CO)OS(=O)(C)=O |
| IUPAC-namn | magnesium;[(2R)-2-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-5-oxo-2H-furan-4-yl] phosphate |
| Molekylvikt (g/mol) | 256.1 |