Läs mer
L-Ascorbic Acid 2-Phosphate Sesquimagnesium Salt Hydrate (~15% Inorganics), TRC
Produktkod. 30347710
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30347710 1 g
1 options
Produktkod. 30347710 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7850101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-Ascorbic Acid 2-Phosphate Sesquimagnesium Salt Hydrate
CAS 1713265-25-8
Molekylformel C6H7MgO9P
Kvantitet 1 g
Synonym L-Ascorbic Acid 2-(Dihydrogen Phosphate) Magnesium Salt Hydrate
InChI formel InChI=1S/C6H9O9P.Mg/c7-1-2(8)4-3(9)5(6(10)14-4)15-16(11,12)13;/h2,4,7-9H,1H2,(H2,11,12,13);/q;+2/p-2/t2-,4+;/m0./s1
LEDER O.OC[C@H](O)[C@H]1OC(=O)C(=C1O)OP(=O)(O)O
IUPAC-namn magnesium;[(2R)-2-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-5-oxo-2H-furan-4-yl] phosphate
Molekylvikt (g/mol) 278.39
Formel vikt 274.009