missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ascorbic acid, Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Appearance | Complying |
Loss on Drying | Max. 0.1 % |
Packaging | Plastic Bottle |
Quantity | 1 kg |
Physical Form | Crystals |
Sulfated Ash | Max. 0.05 % |
Percent Purity | 99.7 - 100.5 % |
Grade | ACS Grade Organics |