missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Ascorbic Acid, Powder, BAKER ANALYZED™ Biochemical Reagent, J.T. Baker™

Catalog Number p-5391067
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
50-81-7
C6H8O6
176.12
MFCD00064328
CIWBSHSKHKDKBQ-DOMZIZNONA-N
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
54670067
CHEBI:29073
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O
This item is not returnable. View return policy
50-81-7
C6H8O6
176.12
MFCD00064328
CIWBSHSKHKDKBQ-DOMZIZNONA-N
l-ascorbic acid, ascorbic acid, vitamin c, l-ascorbate, ascorbate, ascorbicap, l +-ascorbic acid, cevitamic acid, ascoltin, hybrin
54670067
CHEBI:29073
OC[C@H](O)[C@H]1OC(=O)C(O)=C1O

High quality biochemicals for research use

Packaging Poly Bottle
Quantity 100 g
Specific Gravity 1L = 1.65kg
IUPAC Name (2R)-2-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
Formula Weight 176.12
Grade BAKER ANALYZED™ Biochemical Reagent
Physical Form Powder
Chemical Name or Material L-(+)-Ascorbic Acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'