Läs mer
L-Aspartic-13C4 Acid, TRC
Produktkod. 30008313
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008313 5 mg
30008551 10 mg
30010997 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008313 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7902135MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55443-54-4
Molekylformel 13C4H7NO4
Molekylvikt (g/mol) 137.07
Synonym L-Aspartic-1,2,3,4-13C4 acid (ACI), L-Aspartic-13C4 acid (9CI), L-Aspartic Acid-13C4, (2S)-2-Aminobutanedioic-1,2,3,4-13C4 acid
IUPAC-namn (2S)-2-amino(1,2,3,4-13C4)butanedioic acid
LEDER N[13C@@H]([13CH2][13C](=O)O)[13C](=O)O
Specifikationer
Kemiskt namn eller material L-Aspartic-13C4 Acid
Kvantitet 5 mg
InChI formel InChI=1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1/i1+1,2+1,3+1,4+1
Formel vikt 137.0509