Learn More
L-Aspartic acid β-benzyl ester, 98%, Thermo Scientific Chemicals
Catalog Number p-4323639
Click to view available options
Quantity:
5 g
25 g
2177-63-1
C11H13NO4
223.23
VGALFAWDSNRXJK-VIFPVBQESA-N
h-asp obzl-oh, l-aspartic acid 4-benzyl ester, l-aspartic acid beta-benzyl ester, beta-benzyl l-aspartate, 2s-2-amino-4-benzyloxy-4-oxobutanoic acid, 4-benzyl l-aspartate, benzyl hydrogen beta-l-aspartate, s-2-amino-4-benzyloxy-4-oxobutanoic acid, 2s-2-amino-4-oxo-4-phenylmethoxybutanoic acid, asp obzl
101186
(2S)-2-amino-4-oxo-4-phenylmethoxybutanoic acid
C1=CC=C(C=C1)COC(=O)CC(C(=O)O)N
2177-63-1
C11H13NO4
223.23
VGALFAWDSNRXJK-VIFPVBQESA-N
h-asp obzl-oh, l-aspartic acid 4-benzyl ester, l-aspartic acid beta-benzyl ester, beta-benzyl l-aspartate, 2s-2-amino-4-benzyloxy-4-oxobutanoic acid, 4-benzyl l-aspartate, benzyl hydrogen beta-l-aspartate, s-2-amino-4-benzyloxy-4-oxobutanoic acid, 2s-2-amino-4-oxo-4-phenylmethoxybutanoic acid, asp obzl
101186
(2S)-2-amino-4-oxo-4-phenylmethoxybutanoic acid
C1=CC=C(C=C1)COC(=O)CC(C(=O)O)N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 207°C to 208°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Specific Rotation + 27.8
Formula Weight 223.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Aspartic acid β-benzyl ester
Product Identifier
  • L-Aspartic acid beta-benzyl ester