Läs mer
L-Aspartic Acid Calcium Salt Hydrochloride, TRC
Produktkod. 30345119
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30345119 1 g
1 options
Produktkod. 30345119 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7900211G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-Aspartic Acid Calcium Salt Hydrochloride
CAS 92533-40-9
Molekylformel C4H6CaClNO4
Kvantitet 1 g
Synonym L-Aspartic Acid Calcium Salt Hydrochloride, Calcium Aspartate Hydrochloride
InChI formel InChI=1S/C4H7NO4.Ca.ClH/c5-2(4(8)9)1-3(6)7;;/h2H,1,5H2,(H,6,7)(H,8,9);;1H/q;+2;/p-2/t2-;;/m0../s1
LEDER O=C([O-])C[C@H](N)C([O-])=O.[Ca+2].Cl
IUPAC-namn calcium;(2S)-2-aminobutanedioate;hydrochloride
Molekylvikt (g/mol) 207.63
Formel vikt 206.9611