Läs mer
Tocris Bioscience™ L-Azidonorleucine hydrochloride
Beskrivning
L-Azidonorleucine hydrochloride is an unnatural amino acid; surrogate methionine. Can be incorporated into proteins by a mutant methionyl tRNA synthetase to enable cell-type specific analysis of protein synthesis. For use with bio-orthogonal non-canonical amino-acid tagging (BONCAT) or fluorescent non-canonical amino acid tagging (FUNCAT). Labeling does not affect the normal localization of the labeled proteins.
Specifikationer
Specifikationer
| Kemiskt namn eller material | (S)-2-Amino-6-azidohexanoic acid hydrochloride |
| CAS | 1454334-76-9 |
| Kvantitet | 10 mg |
| Molekylformel | C6H12N4O2.HCl |
| Formel vikt | 208.65 |