Learn More
L-Carnitine, 98+%, Thermo Scientific Chemicals
Catalog Number p-7023752
Click to view available options
Quantity:
10 g
50 g
250 g
541-15-1
C7H15NO3
161.201
MFCD00038747
PHIQHXFUZVPYII-ZCFIWIBFSA-N
l-carnitine, levocarnitine, vitamin bt, r-carnitine, carnitor, --carnitine, carnitine, --l-carnitine, karnitin, l---carnitine
10917
CHEBI:16347
(3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate
C[N+](C)(C)CC(CC(=O)[O-])O
541-15-1
C7H15NO3
161.201
MFCD00038747
PHIQHXFUZVPYII-ZCFIWIBFSA-N
l-carnitine, levocarnitine, vitamin bt, r-carnitine, carnitor, --carnitine, carnitine, --l-carnitine, karnitin, l---carnitine
10917
CHEBI:16347
(3R)-3-hydroxy-4-(trimethylazaniumyl)butanoate
C[N+](C)(C)CC(CC(=O)[O-])O
L-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Carnitine is widely used as a nutritional supplement in food and beverage industries. It is used as a food additive. It acts as a cyanide antidote. It is also used in reversing and preventing hyperthyroid symptoms. It is a vitamin-like nutrient, which plays an important role in human metabolism.
Solubility
Soluble in water, pyridine, dimethyl sulfoxide, methanol and ethanol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY
Melting Point ∼190°C (decomposition)
Quantity 10 g
Beilstein 4292315
Sensitivity Hygroscopic
Merck Index 14,1849
Solubility Information Soluble in water,pyridine,dimethyl sulfoxide,methanol and ethanol.
Formula Weight 161.2
Percent Purity ≥99%
Chemical Name or Material L-Carnitine
compliance-icons
Product Identifier
  • L-Carnitine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : (R)-(3-Carboxy-2-hydroxypropyl)trimethyla mmonium hydroxide