Läs mer
L-Cephalexin, TRC
Produktkod. 30353313
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30353313 50 mg
1 options
Produktkod. 30353313 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC25680550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-Cephalexin
CAS 34632-04-7
Molekylformel C16 H17 N3 O4 S
Kvantitet 50 mg
Synonym 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2S)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-, (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[(aminophenylacetyl)amino]-3-methyl-8-oxo-, [6R-[6α,7β(S*)]]-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2S)-aminophenylacetyl]amino]-3-methyl-8-oxo-, (6R,7R)- (9CI), (6R,7R)-7-[[(2S)-2-Amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-(α-Amino-L-phenylacetamido)-3-deacetoxycephalosporanic acid, L-Cephalexin
InChI formel InChI=1S/C16H17N3O4S/c1-8-7-24-15-11(14(21)19(15)12(8)16(22)23)18-13(20)10(17)9-5-3-2-4-6-9/h2-6,10-11,15H,7,17H2,1H3,(H,18,20)(H,22,23)/t10-,11+,15+/m0/s1
LEDER CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@@H](N)c3ccccc3)C2=O)C(=O)O
IUPAC-namn (6R,7R)-7-[[(2S)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molekylvikt (g/mol) 347.39
Formel vikt 347.094