Läs mer
L-(+)-Cystathionine, TRC
Produktkod. 30137000
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137000 50 mg
30136292 5 mg
17989444 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137000 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC98950050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56-88-2
Molekylformel C7 H14 N2 O4 S
Molekylvikt (g/mol) 222.26
Synonym S-[(2R)-2-Amino-2-carboxyethyl]-L-homocysteine, Cystathionine (6CI,7CI), Cystathionine, L- (8CI), L-Homocysteine, S-(2-amino-2-carboxyethyl)-, (R)-, S-[(2R)-2-Amino-2-carboxyethyl]-L-homocysteine, Butanoic acid, 2-amino-4-[(2-amino-2-carboxyethyl)thio]-, [R-(R*,S*)]-, L-(+)-Cystathionine, L-Cystathionine
IUPAC-namn (2S)-2-amino-4-[(2R)-2-amino-2-carboxyethyl]sulfanylbutanoic acid
LEDER N[C@@H](CCSC[C@H](N)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material L-Cystathionine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C7H14N2O4S/c8-4(6(10)11)1-2-14-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13)/t4-,5-/m0/s1
Kvantitet 50 mg
Formel vikt 222.0674
Procent renhet >95%