Läs mer
L-Cysteine Hydrochloride Monohydrate, TRC
Produktkod. 30357131
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30357131 10 g
1 options
Produktkod. 30357131 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC99500510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cysteine hydrochloride monohydrate
CAS 7048-04-6
Molekylformel C3 H7 N O2 S . Cl H . H2 O
Kvantitet 10 g
Synonym Cysteine hydrochloride monohydrate, (2R)-2-amino-3-sulfanylpropanoic acid hydrochloride
InChI formel InChI=1S/C3H7NO2S.ClH.H2O/c4-2(1-7)3(5)6;;/h2,7H,1,4H2,(H,5,6);1H;1H2/t2-;;/m0../s1
LEDER O.Cl.N[C@@H](CS)C(=O)O
IUPAC-namn (2R)-2-amino-3-sulfanylpropanoic acid;hydrate;hydrochloride
Molekylvikt (g/mol) 175.63
Formel vikt 175.007