missing translation for 'onlineSavingsMsg'
Läs mer

L-Cysteinesulfinic Acid, Mikromol™

Produktkod. 30051862
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051862 100 mg
30050787 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30051862 Leverantör Mikromol Leverantörsnummer MM0236.03

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

L-Cysteinesulfinic Acid
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material L-Cysteinesulfinic Acid
CAS 1115-65-7
Analyt- eller komponentnamn L-Cysteinesulfinic Acid
Frakt skick Room Temperature
Molekylformel C3 H7 N O4 S
Certifikat och efterlevnad ISO 17025
Synonym L-Cysteinesulfinic Acid
InChI formel InChI=1S/C3H7NO4S/c4-2(3(5)6)1-9(7)8/h2H,1,4H2,(H,5,6)(H,7,8)/t2-/m0/s1
LEDER N[C@@H](CS(=O)O)C(=O)O
IUPAC-namn (2R)-2-amino-3-sulfinopropanoic acid
Molekylvikt (g/mol) 153.16
Kvantitet 100 mg
Formel vikt 153.0096 g/mol
Visa mer Visa mindre
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.