Läs mer
L-Cysteinyl-L-cysteinyl-L-Alpha-glutamyl-L-tyrosyl-L-cysteinyl-L-cysteinyl-L-asparaginyl-L-prolyl-L-alanyl-L-cysteinyl-L-threonylglycyl-L-cysteinyl-L-Tyrosine Trifluoroacetate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | L-Cysteinyl-L-cysteinyl-L-alpha-glutamyl-L-tyrosyl-L-cysteinyl-L-cysteinyl-L-asparaginyl-L-prolyl-L-alanyl-L-cysteinyl-L-threonylglycyl-L-cysteinyl-L-Tyrosine Trifluoroacetate |
| CAS | 1638632-09-3 |
| Molekylformel | C61H86F3N15O23S6 |
| Kvantitet | 50 mg |
| Synonym | Linaclotide Linear Trifluoroacetate |
| InChI formel | InChI=1S/C59H85N15O21S6.C2HF3O2/c1-26(47(82)69-41(25-101)55(90)73-46(27(2)75)57(92)62-19-44(79)64-37(21-97)51(86)68-36(59(94)95)17-29-7-11-31(77)12-8-29)63-56(91)42-4-3-15-74(42)58(93)35(18-43(61)78)67-53(88)39(23-99)72-54(89)40(24-100)71-50(85)34(16-28-5-9-30(76)10-6-28)66-49(84)33(13-14-45(80)81)65-52(87)38(22-98)70-48(83)32(60)20-96;3-2(4,5)1(6)7/h5-12,26-27,32-42,46,75-77,96-101H,3-4,13-25,60H2,1-2H3,(H2,61,78)(H,62,92)(H,63,91)(H,64,79)(H,65,87)(H,66,84)(H,67,88)(H,68,86)(H,69,82)(H,70,83)(H,71,85)(H,72,89)(H,73,90)(H,80,81)(H,94,95);(H,6,7)/t26-,27+,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,46-;/m0./s1 |
| LEDER | C[C@@H](O)[C@H](NC(=O)[C@H](CS)NC(=O)[C@H](C)NC(=O)[C@@H]1CCCN1C(=O)[C@H](CC(=O)N)NC(=O)[C@H](CS)NC(=O)[C@H](CS)NC(=O)[C@H](Cc2ccc(O)cc2)NC(=O)[C@H](CCC(=O)O)NC(=O)[C@H](CS)NC(=O)[C@@H](N)CS)C(=O)NCC(=O)N[C@@H](CS)C(=O)N[C@@H](Cc3ccc(O)cc3)C(=O)O.OC(=O)C(F)(F)F |
| IUPAC-namn | (4S)-5-[[(2S)-1-[[(2R)-1-[[(2R)-1-[[(2S)-4-amino-1-[(2S)-2-[[(2S)-1-[[(2R)-1-[[(2S,3R)-1-[[2-[[(2R)-1-[[(1S)-1-carboxy-2-(4-hydroxyphenyl)ethyl]amino]-1-oxo-3-sulfanylpropan-2-yl]amino]-2-oxoethyl]amino]-3-hydroxy-1-oxobutan-2-yl]amino]-1-oxo-3-sulfanylpropan-2-yl]amino]-1-oxopropan-2-yl]carbamoyl]pyrrolidin-1-yl]-1,4-dioxobutan-2-yl]amino]-1-oxo-3-sulfanylpropan-2-yl]amino]-1-oxo-3-sulfanylpropan-2-yl]amino]-3-(4-hydroxyphenyl)-1-oxopropan-2-yl]amino]-4-[[(2R)-2-[[(2R)-2-amino-3-sulfanylpropanoyl]amino]-3-sulfanylpropanoyl]amino]-5-oxopentanoic acid;2,2,2-trifluoroacetic acid |
| Molekylvikt (g/mol) | 1646.81 |
| Formel vikt | 1645.4297 |