Läs mer
L-Daunosamine, Hydrochoride, TRC
Produktkod. 30133101
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133101 50 mg
30012437 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133101 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD19475050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 105497-63-0
Molekylformel C6 H13 N O3 . Cl H
Molekylvikt (g/mol) 183.63
Synonym L-Daunosamine HCl
IUPAC-namn (4S,5S,6S)-4-amino-6-methyloxane-2,5-diol;hydrochloride
LEDER Cl.C[C@@H]1OC(O)C[C@H](N)[C@@H]1O
Specifikationer
Kemiskt namn eller material L-Daunosamine, Hydrochoride
Kvantitet 50 mg
InChI formel InChI=1S/C6H13NO3.ClH/c1-3-6(9)4(7)2-5(8)10-3;/h3-6,8-9H,2,7H2,1H3;1H/t3-,4-,5?,6+;/m0./s1
Formel vikt 183.0662