Läs mer
L-α-Dipalmitoylphosphatidyl-N-monomethylethanolamine (>80%), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | L-Alpha-Dipalmitoylphosphatidyl-N-monomethylethanolamine |
| CAS | 3930-13-0 |
| Molekylformel | C38 H76 N O8 P |
| Kvantitet | 2.5 mg |
| Synonym | L-Alpha-Dipalmitoylphosphatidyl-N-monomethylethanolamine |
| InChI formel | InChI=1S/C38H76NO8P/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-37(40)44-34-36(35-46-48(42,43)45-33-32-39-3)47-38(41)31-29-27-25-23-21-19-17-15-13-11-9-7-5-2/h36,39H,4-35H2,1-3H3,(H,42,43)/t36-/m1/s1 |
| LEDER | CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)(O)OCCNC)OC(=O)CCCCCCCCCCCCCCC |
| IUPAC-namn | [(2R)-2-hexadecanoyloxy-3-[hydroxy-[2-(methylamino)ethoxy]phosphoryl]oxypropyl] hexadecanoate |
| Molekylvikt (g/mol) | 705.986 |
| Formel vikt | 705.531 |