Läs mer
L-DOPA Methyl Ester Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | L-DOPA Methyl Ester Hydrochloride |
| CAS | 1421-65-4 |
| Molekylformel | C10 H13 N O4 . Cl H |
| Kvantitet | 1 g |
| Synonym | L-Tyrosine, 3-hydroxy-, methyl ester, hydrochloride (1:1), Alanine, 3-(3,4-dihydroxyphenyl)-, methyl ester, hydrochloride (6CI,7CI), Alanine, 3-(3,4-dihydroxyphenyl)-, methyl ester, hydrochloride, L- (8CI), L-Tyrosine, 3-hydroxy-, methyl ester, hydrochloride (9CI), 3,4-Dihydroxyphenyl-L-alanine methyl ester hydrochloride, L-DOPA Me ester hydrochloride, L-Dopa methyl ester hydrochloride, Levodopa methyl ester hydrochloride, Methyl L-3-(3,4-dihydroxyphenyl)alaninate hydrochloride, ST 41769 |
| InChI formel | InChI=1S/C10H13NO4.ClH/c1-15-10(14)7(11)4-6-2-3-8(12)9(13)5-6;/h2-3,5,7,12-13H,4,11H2,1H3;1H/t7-;/m0./s1 |
| LEDER | Cl.COC(=O)[C@@H](N)Cc1ccc(O)c(O)c1 |
| IUPAC-namn | methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoate;hydrochloride |
| Molekylvikt (g/mol) | 247.68 |
| Formel vikt | 247.0611 |