Läs mer
L-Felinine, TRC
Produktkod. 30240978
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240978 50 mg
30240945 5 mg
30240848 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240978 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF23125050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 471-09-0
Molekylformel C8 H17 N O3 S
Molekylvikt (g/mol) 207.29
IUPAC-namn (2R)-2-amino-3-(4-hydroxy-2-methylbutan-2-yl)sulfanylpropanoic acid
LEDER CC(C)(CCO)SC[C@H](N)C(=O)O
Specifikationer
Kemiskt namn eller material L-Felinine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C8H17NO3S/c1-8(2,3-4-10)13-5-6(9)7(11)12/h6,10H,3-5,9H2,1-2H3,(H,11,12)/t6-/m0/s1
Kvantitet 50 mg
Formel vikt 207.0929
Procent renhet >95%