missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Fenchone, 98+%, Thermo Scientific Chemicals

Catalog Number p-133338
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7787-20-4
C10H16O
152.24
MFCD00151104
LHXDLQBQYFFVNW-GMSGAONNSA-N
l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
3034206
(1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC(C2)(C1=O)C)C
This item is not returnable. View return policy
7787-20-4
C10H16O
152.24
MFCD00151104
LHXDLQBQYFFVNW-GMSGAONNSA-N
l-fenchone, 1r-fenchone, 1r,4r-2,2,4-trimethylbicyclo 2.2.1 heptan-3-one, 2r,5r-2,6,6-trimethyl-2,5-methanocyclohexane-1-one
3034206
(1R,4R)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC(C2)(C1=O)C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 5°C
Color Colorless to Yellow
Density 0.9400g/mL
Boiling Point 192°C to 194°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4610 to 1.463
Quantity 50 g
Fieser 08,228
Specific Gravity 0.94
Specific Rotation Condition −50.50° (24°C neat)
Specific Rotation -50.5
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether
Formula Weight 152.24
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material L(-)-Fenchone, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • L(-)-Fenchone
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'