Läs mer
L-Gamma-Glutamyl-(S)-Allyl-Cysteine, TRC
Produktkod. 17989914
Change view
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17989914 5 mg
17908104 25 mg
17950075 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17989914 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG5970605MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 91216-95-4
Molekylformel C11H18N2O5S
Molekylvikt (g/mol) 290.34
Synonym L-N-[2-(Allylthio)-1-carboxyethyl]glutamine, L-γ-Glutamyl-S-2-propenyl-L-cysteine, N-L-γ-Glutamyl-S-2-propenyl-L-cysteine;
LEDER N[C@@H](CCC(=O)N[C@@H](CSCC=C)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material gamma-Glutamyl-(S)-allyl-L-cysteine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C11H18N2O5S/c1-2-5-19-6-8(11(17)18)13-9(14)4-3-7(12)10(15)16/h2,7-8H,1,3-6,12H2,(H,13,14)(H,15,16)(H,17,18)/t7-,8-/m0/s1
Kvantitet 5 mg
Formel vikt 290.0936
Procent renhet >95%
Frakt skick Dry Pack