Läs mer
L-Glutamine-13C, TRC
Produktkod. 30136197
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30136197 25 mg -
30003859 2.5 mg 2.5mg
30012190 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136197 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG59729825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 159680-32-7
Molekylformel C413CH10N2O3
Molekylvikt (g/mol) 147.14
Synonym L-Glutamine-5-13C (9CI, ACI), L-Glutamine-13C, (2S)-2-Amino-4-(carbamoyl-13C)butanoic acid, (2S)-2,5-Diamino-5-oxopentanoic acid-5-13C
IUPAC-namn (2S)-2,5-diamino-5-oxo(513C)pentanoic acid
LEDER N[C@@H](CC[13C](=O)N)C(=O)O
Specifikationer
Kemiskt namn eller material L-Glutamine-13C
Kvantitet 25 mg
InChI formel InChI=1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1/i4+1
Formel vikt 147.0725