Läs mer
L-Glutamine-13C5, TRC
Produktkod. 30010709
Change view
Kvantitet:
1 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010709 1 mg
30009367 10 mg
30008064 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010709 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG5970011MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 184161-19-1
Molekylformel 13C5H10N2O3
Molekylvikt (g/mol) 151.11
Synonym (S)-2,5-Diamino-5-oxopentanoic Acid-13C5, 2-Aminoglutaramic Acid-13C5, Acustasin-13C5, Aesgen 14-13C5, Cebrogen-13C5, Glumin-13C5, L-2-Aminoglutaramidic Acid-13C5, L-Glutamic Acid γ-αmide-13C5, Levoglutamide-13C5, NSC 27421-13C5, Saforis-13C5, Stimulina-13C5, γ-Glutamine-13C5 ;
IUPAC-namn (2S)-2,5-diamino-5-oxo(1,2,3,4,5-13C5)pentanoic acid
LEDER N[13C@@H]([13CH2][13CH2][13C](N)=O)[13C](O)=O
Specifikationer
Kemiskt namn eller material L-Glutamine-13C5
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1/i1+1,2+1,3+1,4+1,5+1
Formel vikt 151.0859
Procent renhet >95%