Läs mer
L-Glutamine-amide-15N, TRC
Produktkod. 30134840
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134840 100 mg
30133987 250 mg
30006131 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134840 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG597002100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59681-32-2
Molekylformel C5 H10 15N N O3
Molekylvikt (g/mol) 147.14
Synonym L-Glutamine-N-15N (6CI, 9CI, ACI), L-[5-15N]Glutamine, L-Glutamine-amide-15N, L-Glutamine-5-15N, L-Glutamine-15N, (2S)-2-Amino-4-(carbamoyl-15N)butanoic acid, (2S)-2-Amino-5-(amino-15N)-5-oxopentanoic acid
IUPAC-namn (2S)-2-amino-5-(^{15}N)azanyl-5-oxopentanoic acid
LEDER [15NH2]C(=O)CC[C@H](N)C(=O)O
Specifikationer
Kemiskt namn eller material L-Glutamine-amide-15N
InChI formel InChI=1S/C5H10N2O3/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H2,7,8)(H,9,10)/t3-/m0/s1/i7+1
Kvantitet 100 mg
Formel vikt 147.0662