Läs mer
L-Glycero-D-mannoheptose, TRC
Produktkod. 30240801
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240801 10 mg
30240558 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240801 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG59849510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4305-74-2
Molekylformel C7 H14 O7
Molekylvikt (g/mol) 210.18
IUPAC-namn (2S,3S,4R,5R,6S)-2,3,4,5,6,7-hexahydroxyheptanal
Specifikationer
Kemiskt namn eller material L-Glycero-D-mannoheptose
Kvantitet 10 mg
InChI formel InChI=1S/C7H14O7/c8-1-3(10)5(12)7(14)6(13)4(11)2-9/h1,3-7,9-14H,2H2/t3-,4+,5-,6-,7+/m1/s1
LEDER (2S,3S,5R)-3-methyl-4,4,7-trioxo-3-(triazol-1-ylmethyl)-4λ^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Formel vikt 210.074