Läs mer
L-Hercynine, TRC
Produktkod. 30138518
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138518 500 mg
30136942 250 mg
30012621 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138518 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH288900500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 534-30-5
Molekylformel C9 H15 N3 O2
Molekylvikt (g/mol) 197.23
Synonym 1H-Imidazole-4-ethanaminium, α-carboxy-N,N,N-trimethyl-, inner salt, (αS)-, 1H-Imidazole-4-ethanaminium, α-carboxy-N,N,N-trimethyl-, inner salt, (S)-, Ammonium, (1-carboxy-2-imidazol-4-ylethyl)trimethyl-, hydroxide, inner salt, L- (8CI), (2S)-3-(1H-Imidazol-4-yl)-2-(trimethylazaniumyl)propanoate, (2S)-3-(1H-Imidazol-5-yl)-2-(trimethylazaniumyl)propanoate, Histidine-betaine, N-α-trimethyl histidine, α-N,N,N-Trimethylhistidine, L-Hercynine
IUPAC-namn (2S)-3-(1H-imidazol-5-yl)-2-(trimethylazaniumyl)propanoate
LEDER C[N+](C)(C)[C@@H](Cc1cnc[nH]1)C(=O)[O-]
Specifikationer
Kemiskt namn eller material L-Hercynine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C9H15N3O2/c1-12(2,3)8(9(13)14)4-7-5-10-6-11-7/h5-6,8H,4H2,1-3H3,(H-,10,11,13,14)/t8-/m0/s1
Kvantitet 500 mg
Formel vikt 197.1164
Procent renhet >95%