Läs mer
L-Histidine Hydrochloride Hydrate (<5% D) (13C6, 97-99%), TRC
Produktkod. 30133689
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133689 5 mg
30133329 25 mg
30009840 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133689 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH4560405MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 201740-88-7
Molekylformel 13C6H9N3O2 • (HCl) • (H2O)
Molekylvikt (g/mol) 215.58671
Synonym L-Histidine-carboxy,α,β,2,4,5-13C6, hydrochloride, hydrate (1:1:1) (ACI), L-Histidine-13C6, monohydrochloride, monohydrate (9CI), L-Histidine-13C6 monohydrochloride monohydrate, 2-Amino-3-(1H-imidazol-5-yl-2,4,5-13C3)propanoic-1,2,3-13C3 acid monohydrochloride monohydrate
IUPAC-namn (2S)-2-amino-3-((2,4,5-^{13}C_{3})1H-imidazol-4-yl)(1,2,3-^{13}C_{3})propanoic acid;hydrate;hydrochloride
LEDER O.Cl.N[13C@@H]([13CH2][13c]1[13cH][nH][13cH]n1)[13C](=O)O
Specifikationer
Kemiskt namn eller material L-Histidine Hydrochloride Hydrate 13C6
Kvantitet 5 mg
InChI formel InChI=1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1/i1+1,2+1,3+1,4+1,5+1,6+1;;
Formel vikt 215.0768