Läs mer
L-Homocysteine, TRC
Produktkod. 30136886
Change view
Kvantitet:
25 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136886 500 mg
30136641 50 mg
30004725 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136886 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH591950500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6027-13-0
Molekylformel C4 H9 N O2 S
Molekylvikt (g/mol) 135.1848
Synonym L-Homocysteine, Butyric acid, 2-amino-4-mercapto-, L- (8CI), (S)-2-Amino-4-mercaptobutanoic acid, (S)-Homocysteine, 2-Amino-4-mercapto-L-butyric acid, 2-Amino-4-mercaptobutyric acid, Butanoic acid, 2-amino-4-mercapto-, (S)-, Homocysteine, NSC 43117
IUPAC-namn (2S)-2-amino-4-sulfanylbutanoic acid
LEDER N[C@@H](CCS)C(=O)O
Specifikationer
Kemiskt namn eller material L-Homocysteine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C4H9NO2S/c5-3(1-2-8)4(6)7/h3,8H,1-2,5H2,(H,6,7)/t3-/m0/s1
Kvantitet 500 mg
Formel vikt 135.0354
Procent renhet >95%