Läs mer
L-Homocystine, TRC
Produktkod. 30135761
Change view
Kvantitet:
100 mg
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135761 25 g
30133488 10 g
30008636 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135761 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH59136025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 626-72-2
Molekylformel C8 H16 N2 O4 S2
Molekylvikt (g/mol) 268.35
Synonym Butanoic acid, 4,4'-dithiobis[2-amino-, [S-(R*,R*)]-, Butyric acid, 4,4'-dithiobis[2-amino-, L- (8CI), (2S,2'S)-4,4'-Dithiobis[2-aminobutanoic acid], L-Homocystine
IUPAC-namn (2S)-2-amino-4-[[(3S)-3-amino-3-carboxypropyl]disulfanyl]butanoic acid
LEDER N[C@@H](CCSSCC[C@H](N)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material L-Homocystine
Kvantitet 25 g
InChI formel InChI=1S/C8H16N2O4S2/c9-5(7(11)12)1-3-15-16-4-2-6(10)8(13)14/h5-6H,1-4,9-10H2,(H,11,12)(H,13,14)/t5-,6-/m0/s1
Formel vikt 268.0551