Läs mer
L-Homophenylalanine, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | L-Homophenylalanine |
| CAS | 943-73-7 |
| Molekylformel | C10 H13 N O2 |
| Kvantitet | 2.5 g |
| Synonym | Benzenebutanoic acid, α-amino-, (αS)-, Benzenebutanoic acid, α-amino-, (S)-, Butyric acid, 2-amino-4-phenyl-, L- (8CI), (αS)-α-Aminobenzenebutanoic acid, (+)-(S)-Homophenylalanine, (+)-Homophenylalanine, (+)-L-Homophenylalanine, (2S)-2-Amino-4-phenylbutanoic acid, (S)-Homophenylalanine, L-2-Amino-4-phenylbutyric acid, L-Benzylalanine, L-Homophenylalanine, L-γ-Phenylbutyrine, (2S)-2-Amino-4-phenylbutanoic acid |
| InChI formel | InChI=1S/C10H13NO2/c11-9(10(12)13)7-6-8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)/t9-/m0/s1 |
| LEDER | N[C@@H](CCc1ccccc1)C(=O)O |
| IUPAC-namn | (2S)-2-amino-4-phenylbutanoic acid |
| Molekylvikt (g/mol) | 179.22 |
| Formel vikt | 179.0946 |