Läs mer
L-Homopropargylglycine, TRC
Produktkod. 30467962
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30467962 1 mg -
30470878 10 mg -
30480431 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30467962 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL7008001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 98891-36-2
Molekylformel C6 H9 N O2
Molekylvikt (g/mol) 127.14
Synonym (S)-2-Amino-5-hexynoic acid
IUPAC-namn (2S)-2-aminohex-5-ynoic acid
LEDER N[C@@H](CCC#C)C(=O)O
Specifikationer
Kemiskt namn eller material L-Homopropargylglycine
InChI formel InChI=1S/C6H9NO2/c1-2-3-4-5(7)6(8)9/h1,5H,3-4,7H2,(H,8,9)/t5-/m0/s1
Kvantitet 1 mg
Formel vikt 127.0633