Läs mer
L-Iditol, TRC
Produktkod. 30403877
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30403877 100 mg
30403260 50 mg
30408775 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30403877 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI175000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 488-45-9
Molekylformel C6 H14 O6
Molekylvikt (g/mol) 182.17
IUPAC-namn (2S,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
LEDER OC[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO
Specifikationer
Kemiskt namn eller material L-Iditol
Kvantitet 100 mg
InChI formel InChI=1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5+,6+/m0/s1
Formel vikt 182.079