Läs mer
L-idose (0.145M solution), TRC
Produktkod. 30179536
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179536 50 mg
30179576 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179536 Leverantör Toronto Research Chemicals Leverantörsnummer TRCI17506050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5934-56-5
Molekylformel C6 H12 O6
Molekylvikt (g/mol) 180.16
Synonym L-Idose, Idose, L- (8CI)
IUPAC-namn (2R,3S,4R,5S)-2,3,4,5,6-pentahydroxyhexanal
LEDER OC[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O
Specifikationer
Kemiskt namn eller material L-Idose
Kvantitet 50 mg
InChI formel InChI=1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4-,5+,6+/m0/s1
Formel vikt 180.0633
Koncentration eller sammansättning (efter analyt eller komponenter) L-Idose 0.145M