Läs mer
L-Kynurenine, TRC
Produktkod. 30404937
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30404937 50 mg
1 options
Produktkod. 30404937 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK66100550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L-Kynurenine
CAS 2922-83-0
Anteckningar om renhetsgrad HPLC
Molekylformel C10 H12 N2 O3
Kvantitet 50 mg
Synonym Benzenebutanoic acid, α,2-diamino-γ-oxo-, (αS)-, Alanine, 3-anthraniloyl-, L- (8CI), Benzenebutanoic acid, α,2-diamino-γ-oxo-, (S)-, (αS)-α,2-Diamino-γ-oxobenzenebutanoic acid, (2S)-4-(2-Aminophenyl)-2-azaniumyl-4-oxobutanoate, 3-Anthraniloylalanine, Kynurenin, Kynurenine, L-3-(o-Aminobenzoyl)alanine, L-Kynurenine, (S)-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid
InChI formel InChI=1S/C10H12N2O3/c11-7-4-2-1-3-6(7)9(13)5-8(12)10(14)15/h1-4,8H,5,11-12H2,(H,14,15)/t8-/m0/s1
LEDER N[C@@H](CC(=O)c1ccccc1N)C(=O)O
IUPAC-namn (2S)-2-amino-4-(2-aminophenyl)-4-oxobutanoic acid
Molekylvikt (g/mol) 208.21
Formel vikt 208.0848