Läs mer
L,L-Dityrosine Dihydrochloride, TRC
Produktkod. 30377467
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30377467 10 mg
1 options
Produktkod. 30377467 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD49429010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material L,L-Dityrosine Dihydrochloride
CAS 221308-01-6
Anteckningar om renhetsgrad HPLC
Molekylformel C18 H20 N2 O6 . 2 Cl H
Kvantitet 10 mg
Synonym (α3S,α3'S)-α3,α3'-Diamino-6,6'-dihydroxy-[1,1'-biphenyl]-3,3'-dipropanoic Acid Dihydrochloride, [S-(R*,R*)]- α,α'-Diamino-6,6'-dihydroxy-[1,1'-biphenyl]-3,3'-
InChI formel InChI=1S/C18H20N2O6.2ClH/c19-13(17(23)24)7-9-1-3-15(21)11(5-9)12-6-10(2-4-16(12)22)8-14(20)18(25)26;;/h1-6,13-14,21-22H,7-8,19-20H2,(H,23,24)(H,25,26);2*1H/t13-,14-;;/m0../s1
LEDER Cl.Cl.N[C@@H](Cc1ccc(O)c(c1)c2cc(C[C@H](N)C(=O)O)ccc2O)C(=O)O
IUPAC-namn (2S)-2-amino-3-[3-[5-[(2S)-2-amino-2-carboxyethyl]-2-hydroxyphenyl]-4-hydroxyphenyl]propanoic acid;dihydrochloride
Molekylvikt (g/mol) 433.28
Formel vikt 432.0855